missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14346-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Deubiquitinating enzymes (DUBs) are capable of producing a free ubiquitin moiety from ubiquitin-fused precursors and ubiquitinylated proteins in cells. DUBs possess conserved catalytic regions known as the ′cys domain′ and the ′his domain.′ There are 2 distinct families of DUBs: ubiquitin-specific processing proteases (UBPs, or USPs) and ubiquitin C-terminal hydrolases (UCHs).Specifications
USP1 | |
Polyclonal | |
Unconjugated | |
USP1 | |
Deubiquitinating enzyme 1, hUBP, ubiquitin specific peptidase 1, Ubiquitin thiolesterase 1, UBP, USP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
230484, 313387, 7398 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O94782, Q569C3, Q8BJQ2 | |
Usp1 | |
USP1 Fusion Protein Ag5686 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |