missing translation for 'onlineSavingsMsg'
Learn More

USP16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14055-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16822524

  • 3313.85 SEK / 150µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a deubiquitinating enzyme that is phosphorylated at the onset of mitosis and then dephosphorylated at the metaphase/anaphase transition. It can deubiquitinate H2A, one of two major ubiquitinated proteins of chromatin, in vitro and a mutant form of the protein was shown to block cell division. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications

Specifications

USP16
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q2KJ09, Q99LG0, Q9Y5T5
Deubiquitinating enzyme 16, Ubiquitin thiolesterase 16, UBP M, USP16
Rabbit
IgG
150 μL
-20°C
Primary
10600, 288306, 74112
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
USP16
Liquid
Usp16
USP16 Fusion Protein Ag4981
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers