missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP18 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | USP18 |
---|---|
Concentration | 0.2 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16839693
|
Proteintech
12153-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16829693
|
Proteintech
12153-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
USP18, a member of the deubiquitinating protease family of enzymes, removes ubiquitin adducts from a broad range of protein substrates.Specifications
USP18 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9UMW8 | |
11274 | |
USP18 Fusion Protein Ag2785 | |
Primary | |
-20°C | |
USP18 |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
43 kDa ISG15 specific protease, hUBP43, ISG43, Ubl thiolesterase 18, UBP43, USP18 | |
USP18 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |