missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP8 Mouse anti-Human, Unconjugated, Clone: 1E12E3, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | USP8 |
---|---|
Clone | 1E12E3 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16849885
|
Proteintech
67321-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16839885
|
Proteintech
67321-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The USP8 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human USP8. This antibody recognizes human, mouse antigen. The USP8 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
USP8 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
9101 | |
Primary |
1E12E3 | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human, Mouse | |
IgG1 |