missing translation for 'onlineSavingsMsg'
Learn More

Uteroglobin Rabbit anti-Human, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26909-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16825245

  • 1236.20 SEK / 20µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Uteroglobin encodes a member of the secretoglobin family of small secreted proteins. The encoded protein has been implicated in numerous functions including anti-inflammation, inhibition of phospholipase A2 and the sequestering of hydrophobic ligands. Defects in this gene are associated with a susceptibility to asthma.
Specifications

Specifications

Uteroglobin
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P11684, P17559
CC10, CC16, CCPBP, CCSP, SCGB1A1, UGB, UP 1, UP1, Urinary protein 1, Urine protein 1, Uteroglobin
Rabbit
IgG
20 μL
-20°C
Primary
25575, 7356
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
Unconjugated
SCGB1A1
Liquid
SCGB1A1
Uteroglobin Fusion Protein Ag25530
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers