missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ UTS2 Polyclonal Antibody
GREENER_CHOICE

Produktkod. 16639194
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
16639194 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16639194 Supplier Invitrogen™ Supplier No. PA5102735

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total UTS2.

Urotensin II is an active cyclic heptapeptide absolutely conserved from lamprey to human. The active peptide acts as a vasoconstrictor and is expressed only in brain tissue. Despite the gene family name similarity, this gene is not homologous to urocortin, a member of the sauvagine/corticotropin releasing factor/urotensin I family. Most of the proprotein is cleaved to make the mature peptide.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UTS2
Användningsområden Immunohistochemistry (Paraffin), Western Blot
Klassificering Polyclonal
Koncentration 1 mg/mL
Konjugera Unconjugated
Formulering PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gen UTS2
Genaccessionsnr. O95399, Q9QZQ4
Gene Alias prepro U-II; prepro-UII; PRO1068; Ucn2; UII; U-II; UNQ525/PRO1068; urocortin 2; urotensin 2; urotensin II; Urotensin-2; urotensin-II; UTS2
Gensymboler UTS2
Värd art Rabbit
Immunogen A synthesized peptide derived from human UTS2(Accession O95399), corresponding to amino acid residues Q88-V124.
Reningsmetod Affinity chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 10911, 29180
Målarter Human, Rat
Innehåll och lagring -20°C
Produkttyp Antibody
Form Liquid
Isotyp IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.