missing translation for 'onlineSavingsMsg'
Learn More

Biochrom UV Grade Silica Standard Cuvette with Stopper

Standard rectangular cell with stopper for visible/UV spectrophotometry. 2 mL minimum working volume.

Brand:  Biochrom 80-2002-70

Additional Details : Weight : 0.01700kg

Product Code. 11380132

  • 3755.00 SEK / Each
Estimated Shipment: 23-07-2024
to see stock.
Add to basket

Description

Description

  • Standard rectangular cell with stopper for visible and UV spectrophotometry
  • Minimum working volume 2 mL
  • 10 mm pathlength
  • 12.5 mm x 48 mm external dimensions
  • 10 mm internal width
  • Fits standard spectrophotometer cell holders
Specifications

Specifications

Type 1 Standard Cuvette
UV Grade Silica
10 mm
10 mm
48 mm
Rectangle
2000 μL
12.5 mm
1 Ea.
Dual Beam Ultrospec Spectrophotometers
Spectrophotometric Measurements
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers