Learn More
Biochrom Quartz Matched Micro Cuvette
10 mm Quartz Micro Cell with lid and black walls. 10 mm pathlength with 400 μL minimum volume.
Brand:  Biochrom 80-2109-83
Additional Details : Weight : 0.30000kg
Product Code. 11759545
  • 67835.00 SEK / Pack of 8
Estimated Shipment: 24-07-2024
to see stock.
Add to basket
Description
Description
  • 10 mm Quartz Micro Cell with lid and black walls
  • 10 mm pathlength
  • Minimum volume: 400 μL
  • Fits standard spectrophotometer cell holders
  • For use with Ultrospec 7000/8000/9000, Ultrospec 2100 Pro, GeneQuant 1300/100
Specifications
Specifications
Type 8 Micro Cuvette
UV Grade Silica
8 Pk.
Dual Beam Ultrospec Spectrophotometers
400 μL
2 mm
10 mm
Spectrophotometric Measurements