missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ Black Glasses Case
114.68 SEK valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Use promo code "21673" to get your promotional price.
Click to view available options
Color:
Black
Specifications
Description | Glasses Case |
Color | Black |
Product Type | Eyewear case |
For Use With (Application) | For eyewear |
No. per Pack | 1 Ea. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'