missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ SECU Box
165.07 SEK valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Use promo code "21673" to get your promotional price.
Click to view available options
Color: Blue and red
Description
Specifications
Color | Blue, red |
Product Type | Eyewear storage box |
For Use With (Equipment) | Uvex™ Goggles |
No. per Pack | 1 Ea. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'