missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ V-ATPase G3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555953
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: AKEEAMVEID QYRMQRDKEF RLKQSKIMGS QNNLSDEIEE QTLGKIQELN GHYNKYMESV MNQLLSMVCD MKPEIHVNYR Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 78%.
ATP6V1G3 (V-ATPase G3) is a catalytic subunit of the peripheral V1 complex of vacuolar ATPase (V-ATPase). V-ATPase is responsible for acidifying a variety of intracellular compartments in eukaryotic cells.
Specifikationer
| V-ATPase G3 | |
| Polyclonal | |
| Unconjugated | |
| ATP6V1G3 | |
| ATP6G3; ATP6V1G3; ATPase H+ transporting V1 subunit G3; ATPase, H+ transporting, lysosomal (vacuolar proton pump) subunit G3; ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G3; ATPase, H+ transporting, lysosomal V1 subunit G3; ATPase, H+ transporting, lysosomal, V1 subunit G3; ATPase, H+ transporting, V1 subunit G, isoform 3; vacuolar ATP synthase subunit G 3; vacuolar proton pump G subunit 3; vacuolar proton pump subunit G 3; vacuolar proton pump, subunit G3; V-ATPase 13 kDa subunit 3; V-ATPase G subunit 3; V-ATPase G3 subunit; V-ATPase subunit G 3; Vma10; V-type proton ATPase subunit G 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 127124 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96LB4 | |
| ATP6V1G3 | |
| Recombinant protein corresponding to Human V-ATPase G3. Recombinant protein control fragment (Product #RP-94314). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering