missing translation for 'onlineSavingsMsg'
Learn More

Systec™ V-Series Vertical Floor-Standing Autoclaves, Systec VB

Sterilize solids, liquids and hazardous biological substances with these autoclaves. Systec™ V-Series Vertical Floor-Standing Autoclaves feature a compact and space-saving design with increased chamber height. For simple process applications.

Brand:  Systec™ 1400

Additional Details : Weight : 95.00000kg

 View more versions of this product

Product Code. 12072979

  • 85773.00 SEK / Each


Estimated Shipment: 24-11-2021
Add to basket

Description

Description

Systec VB Top-loading Vertical Autoclave
  • Internal heating elements within the autoclave chamber
  • Housing, support frame and pressure vessel made of corrosion-resistant stainless steel
  • Temperature and pressure range 140°C, 4 bar
  • LCD display and fully automatic microprocessor control
  • 3 sterilization programs
  • Code-secured access rights for changing parameters and further safety-relevant intervention
  • Flexible PT-100 temperature sensor
  • Optional integrated printer for batch documentation
Specifications

Specifications

740mm
450mm
For all laboratory applications even for sophisticated state-of-the-art sterilization processes
VB-40
140°C
LCD display and fully automatic microprocessor control
4 bar
500mm (external)
Top-loading
344mm
920mm (external)
Vertical floor-standing autoclave
V-Series Vertical Floor-Standing Autoclave
220 to 240V, 50/60Hz, 16A
45L (total); 40L (nominal)
Videos
SDS
Documents
Certificates
View accessory products