missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Systec™ V-Series Vertical Floor-Standing Autoclaves, Systec VB
Sterilize solids, liquids and hazardous biological substances with these autoclaves. Systec™ V-Series Vertical Floor-Standing Autoclaves feature a compact and space-saving design with increased chamber height. For simple process applications.
POA
Specifications
Design | Top-loading |
---|---|
For Use With (Application) | For all laboratory applications even for sophisticated state-of-the-art sterilization processes |
Product Type | Autoclave, Vertical |
Temperature (Metric) | 140°C |
Display | LCD display and fully automatic microprocessor control |
Description
Systec VB Top-loading Vertical Autoclave- Internal heating elements within the autoclave chamber
- Housing, support frame and pressure vessel made of corrosion-resistant stainless steel
- Temperature and pressure range 140°C, 4 bar
- LCD display and fully automatic microprocessor control
- 3 sterilization programs
- Code-secured access rights for changing parameters and further safety-relevant intervention
- Flexible PT-100 temperature sensor
- Optional integrated printer for batch documentation
Specifications
Top-loading | |
Autoclave, Vertical | |
LCD display and fully automatic microprocessor control |
For all laboratory applications even for sophisticated state-of-the-art sterilization processes | |
140°C | |
4 bar |