Learn More
Systec™ V-Series Vertical Floor-Standing Autoclaves, Systec VE
Sterilize solids, liquids and hazardous biological substances with these autoclaves. Systec™ V-Series Vertical Floor-Standing Autoclaves feature a compact and space-saving design with increased chamber height for up to 50% more loading capacity. Ideal for basic laboratory applications and media sterilization.
Brand: Systec™ 1255
Description
Systec VE Vertical Top-loading AutoclaveStandard Features
- Internal heating elements within the autoclave chamber
- Housing, support frame and pressure vessel made of corrosion-resistant stainless steel
- Temperature and pressure range 140°C, 4 bar
- LCD display and fully automatic microprocessor control
- 12 sterilization programs
- Code-secured access rights for changing parameters and further safety-relevant intervention
- Internal memory for storing up to 500 sterilization cycles
- Timer for starting programs
- Autofill: automatic demineralized water feed for steam generation
- Flexible PT-100 temperature sensor
- Temperature holding function for liquids after program finish
- Special program for Durham tubes
- Calculation of F0 value
- Special program for waste sterilization with pulsed heat-up for more efficient air exhaust
- Water-cooled steam exhaust, thermostatically controlled
- Programmable automatic door-opening on completion of program
- RS-232 and RS-485 interfaces for external data transmission (network-compatible)
- Extension of temperature and pressure ranges to 150°C / 5 bar (from chamber volume 65 liters and more)
- Rapid cooling for efficient and safe cooling of liquids
- Exhaust filtration (including condensate inactivation) for safe sterilization of hazardous biological substances
- Options for documentation
- Integrated printer for batch documentation
- PC software for comprehensive documentation
- Documentation SD: data storage on SD card for up to 10,000 sterilization cycles and transmission of data to a PC
Specifications
740 mm | |
60 L (Total), 55 L (Nominal) | |
344 mm | |
920 mm | |
Autoclave, Vertical | |
LCD display and fully automatic microprocessor control | |
4 bar |
Top-loading | |
600 mm | |
Basic laboratory applications and media sterilization | |
E-55 | |
140°C | |
220/240 V 50/60 Hz 16 A | |
500 mm |