Learn More
Buerkle™ V4A Stainless Steel PharmaScoop
Buerkle™ V4A Stainless Steel PharmaScoop™ is made without edges, crevices or undercuts, to avoid cross-contamination.
2360.00 SEK - 2895.00 SEK
Product Code | Brand | Capacity (Metric) | Dimensions (L x W) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Dimensions (L x W) | Price | Quantity & Availability | ||||
10263652
|
Buerkle™
5324-0016 |
50 mL | 38 x 70 mm |
2895.00 SEK
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
10355522
|
Buerkle™
5324-0012 |
100 mL | 100 x 50 mm |
2360.00 SEK
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
11911561
|
Buerkle™
5324-0018 |
500 mL | 140 x 95 mm |
2370.00 SEK
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
10435762
|
Buerkle™
5324-0013 |
750 mL | 210 x 95 mm |
2440.00 SEK
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
10204642
|
Buerkle™
5324-0014 |
1100 mL | 235 x 110 mm |
2700.00 SEK
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
10545904
|
Buerkle™
5324-0015 |
1250 mL | 195 x 125 mm |
2785.00 SEK
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
Description
- Stainless steel AISI 316 (1.4401)
- GMP conform
- Highly polished
- Highly polished inner and outer surface