missing translation for 'onlineSavingsMsg'
Learn More
Learn More
V5 Epitope Tag Antibody (SV5-P-K), DyLight 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-81037G
This item is not returnable.
View return policy
Description
V5 Epitope Tag Monoclonal antibody specifically detects V5 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen).Specifications
V5 Epitope Tag | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Rat | |
Protein A purified | |
RUO | |
Recognises a 14 amino acid sequence (GKPIPNPLLGLDST) derived from the P and V proteins of the paramyxoVirus simian virus 5. | |
Purified | |
Epitope Tag |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
SV5-P-K | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
V5 epitope tag | |
Pk (V5) Epitope Tag (GKPIPNPLLGLDST). | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |