missing translation for 'onlineSavingsMsg'
Learn More
Learn More
V5 Epitope Tag Antibody (SV5-P-K), Janelia Fluor™ 549, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP2-81035JF549
This item is not returnable.
View return policy
Description
V5 Epitope Tag Monoclonal antibody specifically detects V5 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen).Specifications
V5 Epitope Tag | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Human | |
Protein A purified | |
RUO | |
Recognises a 14 amino acid sequence (GKPIPNPLLGLDST) derived from the P and V proteins of the paramyxoVirus simian virus 5. | |
Purified | |
Epitope Tag |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
SV5-P-K | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
V5 epitope tag | |
Pk (V5) Epitope Tag (GKPIPNPLLGLDST). | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |