Läs mer
Vaccenic Acid Ethyl Ester, TRC
Produktkod. 30449784
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30449784 250 mg
30455482 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30449784 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV070005250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Vaccenic Acid Ethyl Ester
CAS 89296-48-0
Molekylformel C20H38O2
Kvantitet 250 mg
Synonym 11-Octadecenoic Acid Ethyl Ester, (E)-11-Octadecenoic Acid Ethyl Ester, Ethyl Vaccenate
InChI formel InChI=1S/C20H38O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h9-10H,3-8,11-19H2,1-2H3/b10-9+
LEDER O=C(OCC)CCCCCCCCC/C=C/CCCCCC
IUPAC-namn ethyl (E)-octadec-11-enoate
Molekylvikt (g/mol) 310.51
Formel vikt 310.29