Läs mer
Vaccenic Acid, TRC
Produktkod. 30455105
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30455105 250 mg
1 options
Produktkod. 30455105 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV070000250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material trans-Vaccenic Acid
CAS 693-72-1
Molekylformel C18 H34 O2
Kvantitet 250 mg
Synonym 11-Octadecenoic acid','(E)- (8CI), Vaccenic acid (6CI), (11E)-11-Octadecenoic acid, (E)-11-Octadecenoic acid, (E)-Octadec-11-enoic acid, 11(E)-Octadecenoic acid, 11-trans-Octadecenoic acid, trans-11-Octadecenoic acid, trans-Vaccenic acid, trans-Δ11-Octadecenoic acid, trans-11-Octadecanoic Acid, 11-Octadecenoic acid','(11E)-
InChI formel InChI=1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h7-8H,2-6,9-17H2,1H3,(H,19,20)/b8-7+
LEDER CCCCCC\C=C\CCCCCCCCCC(=O)O
IUPAC-namn (E)-octadec-11-enoic acid
Molekylvikt (g/mol) 282.46
Formel vikt 282.2559