Learn More
BD Vacutainer™ Flashback Blood Collection Needles
Provides instant confirmation of needle placement through a visual flashback system. BD Vacutainer™ Flashback Blood Collection Needles are sterile, multi-sample needles with a safety valve and pre-attached BD tube holder. Available in several lengths and gauges.
POA
Specifications
Product Type | Blood Collection Needle |
---|---|
Sterility | Sterile |
Connectors | Vacutainer |
For Use With (Application) | Blood Collection |
For Use With (Equipment) | Complements the entire Vacutainer range of blood collection devices and accessories |
Description
The BD Vacutainer™ system is optimally designed and tested for controlling pre-analytical variability throughout the specimen collection and handling process.- Provides instant confirmation of needle placement through a unique visual flashback indication
- Avoids the need to probe for a vein or to stick a patient multiple times when the vein is hard to find
- Ensures easy training of personnel and full utilization of closed system collection
- Multi-sample needle ideally complements the range of Vacutainer blood collection line
Specifications
Blood Collection Needle | |
Vacutainer | |
Complements the entire Vacutainer range of blood collection devices and accessories |
Sterile | |
Blood Collection |