Learn More
Merck Millipore Vacuum Filtering Side-Arm Flask
Designed for use in vacuum filtration procedures with classic Glass Filter Holders
638.00 SEK - 2893.00 SEK
Specifications
For Use With (Application) Vacuum Filtration Using Classic Filter Holders
Material Borosilicate glass
View More Specs
Products 3
Product Code Brand Capacity (Metric) Description Price Quantity & Availability  
Product Code Brand Capacity (Metric) Description Price Quantity & Availability  
15875654
View Documents
Merck Millipore
XX1014705
1 L Threaded side-arm 638.00 SEK
Each
Qty Price Break Available View alternative products
Save Up to 
»
Estimated Shipment: 30-06-2021
Add to basket
15884166
View Documents
Merck Millipore
XX1514706
1 L Threaded side-arm with quick vacuum disconnect
934.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 30-06-2021
Add to basket
15810575
View Documents
Merck Millipore
XX1014744
4 L Threaded side-arm
2893.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-06-2021
Add to basket
Description
Description
Designed for use in vacuum filtration procedures with classic Glass Filter Holders
  • Side arm connects to vacuum source with 3/8 in. I.D. hose
  • 1 L and 4 L flasks accept No. 8 perforated stopper
  • 125 mL flask accepts No. 5 stopper
Specifications
Specifications
Vacuum Filtration Using Classic Filter Holders
Borosilicate glass