Learn More
TPP™ Vacuum Filtration “rapid”- Filtermax
Large quadratic filter surface. TPP Vacuum Filtration “rapid”- Filtermax features premium PES filter membrane, pore size of 0.22μm.
1330.30 SEK - 3296.60 SEK
Specifications
Material | Polystyrene, PES |
---|---|
Material (Membrane) | PES |
Pore Size | 0.22 μm |
Type | Filtration Unit |
Product Code | Brand | Description | Filtration Area | No. per Case | Volume (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Filtration Area | No. per Case | Volume (Metric) | Price | Quantity & Availability | ||
11391794
|
TPP™
99155 |
Dimensions: 90 x 89 x 57 mm | 49 cm2 | 36 | 150 mL |
3296.60 SEK
Pack of 36 |
Estimated Shipment: 29-04-2024
Log in to see stock. |
||
11311804
|
TPP™
99255 |
Dimensions: 90 x 89 x 75 mm | 49 cm2 | 24 | 250 mL |
1330.30 SEK
Pack of 24 |
Estimated Shipment: 29-04-2024
Log in to see stock. |
||
11371804
|
TPP™
99955 |
Dimensions: 108 x 108 x 143 mm | 69 cm2 | 12 | 1000 mL |
1743.70 SEK
Pack of 12 |
Estimated Shipment: 26-04-2024
Log in to see stock. |
||
11381794
|
TPP™
99150 |
Dimensions: 90 x 89 x 57 mm | 49 cm2 | 18 | 150 mL |
1727.80 SEK
Pack of 18 |
Estimated Shipment: 29-04-2024
Log in to see stock. |
||
15208734
|
TPP™
99950 |
Dimensions: 111 x 111 x 285 mm | 69 cm2 | 9 | 1000 mL |
2284.30 SEK
Pack of 9 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||
Description
Large quadratic filter surface. TPP Vacuum Filtration “rapid”- Filtermax features premium PES filter membrane, pore size of 0.22μm.
- Large quadratic filter surface
- High efficient PES filter membrane
- Pore size: 0.22μm
- Low protein binding
- High flow rate
- High chemical resistance
Specifications
Polystyrene, PES | |
PES | |
0.22 μm | |
Filtration Unit |