Learn More
Lenz Laborglasintrumente Vacuum Stopcocks, 1-Way, Parallel
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 02942910
Additional Details : Weight : 0.50000kg
Description
- Acc. to DIN 12545 and in addition to DIN standard
- With parallel side arms and hollow glass plug
- Material: borosilicate glass 3.3
- Leakage rate acc. to DIN 12540-1
Specifications
10 mm | |
Borosilicate Glass (3.3) |