Läs mer
Valdecoxib, MedChemExpress
Produktkod. 16442877
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
Förpackningsstorlek:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16442877 50 mg 50mg
16426808 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16442877 Leverantör Medchem Express Leverantörsnummer HY15762
MedChemExpress Valdecoxib is a highly potent and selective inhibitor of COX-2, with IC50s of 5 nM and 140 μM for COX-2 and COX-1, respeceively. Valdecoxib can be used in the research of arthritis and pain.
Valdecoxib is a highly potent and selective inhibitor of COX-2, with IC50s of 5 nM and 140 μM for COX-2 and COX-1, respeceively. Valdecoxib can be used in the research of arthritis and pain.
Specifikationer
Kemiskt namn eller material Valdecoxib
CAS 181695-72-7
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C16H14N2O3S
Kvantitet 50 mg
Synonym SC 65872
Löslighetsinformation DMSO : ≥ 34 mg/mL (108.16 mM)
LEDER O=S(C1=CC=C(C2=C(C)ON=C2C3=CC=CC=C3)C=C1)(N)=O
Molekylvikt (g/mol) 314.36
Formel vikt 314.36
Procent renhet 99.42%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation