Läs mer
Tocris Bioscience™ Valdecoxib
GREENER_CHOICE
Produktkod. 18055542
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18055542 50 mg
18023932 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18055542 Leverantör Tocris Bioscience™ Leverantörsnummer 4206/50
Selective and potent COX-2 inhibitor
Selective and potent COX-2 inhibitor (in vitro IC50 values are 0.005 and 140 μM for human recombinant COX-2 and COX-1 respectively). Displays potent anti-inflammatory activity in vivo.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 181695-72-7
Molekylformel C16H14N2O3S
Specifikationer
Kemiskt namn eller material Valdecoxib
Kvantitet 50 mg
Formel vikt 314.36
Renhet >99%
Molekylvikt (g/mol) 314.359