Läs mer
Valeric Acid Sodium Salt, TRC
Produktkod. 30454027
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30454027 500 mg
1 options
Produktkod. 30454027 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV091420500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Valeric acid sodium salt
CAS 6106-41-8
Molekylformel C5 H9 O2 . Na
Kvantitet 500 mg
Synonym Pentanoic acid','sodium salt (1:1), Pentanoic acid','sodium salt (9CI), Valeric acid','sodium salt (8CI), Sodium n-pentanoate, Sodium n-valerate, Sodium pentanoate, Sodium valerate
InChI formel InChI=1S/C5H10O2.Na/c1-2-3-4-5(6)7;/h2-4H2,1H3,(H,6,7);/q;+1/p-1
LEDER [Na+].CCCCC(=O)[O-]
IUPAC-namn sodium;pentanoate
Molekylvikt (g/mol) 124.11
Formel vikt 124.05