missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Valganciclovir Hydrochloride, Mikromol™
Beskrivning
Valganciclovir Hydrochloride
Specifikationer
Specifikationer
| Kemiskt namn eller material | Valganciclovir Hydrochloride |
| CAS | 175865-59-5 |
| Analyt- eller komponentnamn | Valganciclovir Hydrochloride |
| Frakt skick | Ice pack (-20°C) |
| Molekylformel | C14 H22 N6 O5 . Cl H |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | L-Valine, 2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]-3-hydroxypropyl ester, hydrochloride (1:1), L-Valine, 2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]-3-hydroxypropyl ester, monohydrochloride (9CI), RS 079070-194, Ro 107-9070/194, Valcyte, Valganciclovir hydrochloride |
| InChI formel | InChI=1S/C14H22N6O5.ClH/c1-7(2)9(15)13(23)24-4-8(3-21)25-6-20-5-17-10-11(20)18-14(16)19-12(10)22;/h5,7-9,21H,3-4,6,15H2,1-2H3,(H3,16,18,19,22);1H/t8?,9-;/m0./s1 |
| LEDER | Cl.CC(C)[C@H](N)C(=O)OCC(CO)OCn1cnc2C(=O)N=C(N)Nc12 |
| IUPAC-namn | [2-[(2-amino-6-oxo-3H-purin-9-yl)methoxy]-3-hydroxypropyl] (2S)-2-amino-3-methylbutanoate;hydrochloride |
| Visa mer |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?