Läs mer
Valnemulin Hydrochloride, TRC
Produktkod. 30003979
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30003979 50 mg
30004459 10 mg
30006750 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30003979 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV09440050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 133868-46-9
Molekylvikt (g/mol) 601.28
Synonym (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5-hydroxy-4,6,9,10-tetramethyl-1-oxodecahydro-3a,9-propano-3aH-cyclopenta[8]annulen-8-yl [[2-[[(2R)-2-amino-3-methylbutanoyl]amino]-1,1-dimethylethyl]sulfanyl]acetate hydrochloride, Acetic acid, 2-[[2-[[(2R)-2-amino-3-methyl-1-oxobutyl]amino]-1,1-dimethylethyl]thio]-, (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-ethenyldecahydro-5-hydroxy-4,6,9,10-tetramethyl-1-oxo-3a,9-propano-3aH-cyclopentacycloocten-8-yl ester, hydrochloride (1:1), Acetic acid, [[2-[(2-amino-3-methyl-1-oxobutyl)amino]-1,1-dimethylethyl]thio]-, 6-ethenyldecahydro-5-hydroxy-4,6,9,10-tetramethyl-1-oxo-3a,9-propano-3aH-cyclopentacycloocten-8-yl ester, monohydrochloride, [3aS-[3aα,4β,5α,6α,8β(S*),9α,9aβ,10S*]]-, Acetic acid, [[2-[[(2R)-2-amino-3-methyl-1-oxobutyl]amino]-1,1-dimethylethyl]thio]-, (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-ethenyldecahydro-5-hydroxy-4,6,9,10-tetramethyl-1-oxo-3a,9-propano-3aH-cyclopentacycloocten-8-yl ester, monohydrochloride (9CI), 3a,9-Propano-3aH-cyclopentacyclooctene, acetic acid deriv., Valnemulin hydrochloride
IUPAC-namn (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5-hydroxy-4,6,9,10-tetramethyl-1-oxodecahydro-3a,9-propano-3aH-cyclopenta[8]annulen-8-yl [[2-[[(2R)-2-amino-3-methylbutanoyl]amino]-1,1-dimethylethyl]sulfanyl]acetate hydrochloride
LEDER Cl.CC(C)[C@@H](N)C(=O)NCC(C)(C)SCC(=O)O[C@@H]1C[C@@](C)(C=C)[C@@H](O)[C@H](C)[C@@]23CC[C@@H](C)[C@]1(C)[C@@H]2C(=O)CC3
Specifikationer
Kemiskt namn eller material Valnemulin hydrochloride
Anteckningar om renhetsgrad HPLC
Molekylformel C31 H52 N2 O5 S . Cl H
Kvantitet 50 mg
InChI formel InChI=1S/C31H52N2O5S.ClH/c1-10-29(8)15-22(38-23(35)16-39-28(6,7)17-33-27(37)24(32)18(2)3)30(9)19(4)11-13-31(20(5)26(29)36)14-12-21(34)25(30)31;/h10,18-20,22,24-26,36H,1,11-17,32H2,2-9H3,(H,33,37);1H/t19-,20+,22-,24-,25+,26+,29-,30+,31+;/m1./s1
Formel vikt 600.3364
Procent renhet >95%