Läs mer
Valproic Acid Ethyl Ester, TRC
Produktkod. 30454340
Change view
Kvantitet:
500 mg
2.5 g
5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30454340 500 mg -
30450052 5 g -
30454237 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30454340 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV094770500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17022-31-0
Molekylformel C10 H20 O2
Molekylvikt (g/mol) 172.26
Synonym 2-Propylpentanoic acid ethyl ester, Valeric acid, 2-propyl-, ethyl ester (7CI,8CI), 2-Propylvaleric acid ethyl ester, Ethyl 2,2-dipropylacetate, Ethyl 2-propylpentanoate, Ethyl dipropylacetate, Ethyl valproate, Valproic acid ethyl ester
IUPAC-namn ethyl 2-propylpentanoate
LEDER CCCC(CCC)C(=O)OCC
Specifikationer
Kemiskt namn eller material Ethyl Valproate
Kvantitet 500 mg
InChI formel InChI=1S/C10H20O2/c1-4-7-9(8-5-2)10(11)12-6-3/h9H,4-8H2,1-3H3
Formel vikt 172.1463