Läs mer
Valsartan Benzyl Ester, TRC
Produktkod. 30011589
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011589 10 mg
30004482 5 mg
30011126 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011589 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV09576010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 137863-20-8
Molekylformel C31 H35 N5 O3
Molekylvikt (g/mol) 525.64
Synonym Benzyl (2S)-3-Methyl-2-[pentanoyl-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]amino]butanoate, Valsartan Imp B (EP), Valsartan Related Compound C (USP)
IUPAC-namn benzyl (2S)-3-methyl-2-[pentanoyl-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]amino]butanoate
LEDER CCCCC(=O)N(Cc1ccc(cc1)c2ccccc2c3nnn[nH]3)[C@@H](C(C)C)C(=O)OCc4ccccc4
Specifikationer
Kemiskt namn eller material N-valeryl-N-{[2'-(1H-tetrazole-5-yl)biphenyl-4-yl]methyl}-L-valine benzyl ester
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C31H35N5O3/c1-4-5-15-28(37)36(29(22(2)3)31(38)39-21-24-11-7-6-8-12-24)20-23-16-18-25(19-17-23)26-13-9-10-14-27(26)30-32-34-35-33-30/h6-14,16-19,22,29H,4-5,15,20-21H2,1-3H3,(H,32,33,34,35)/t29-/m0/s1
Formel vikt 525.274
Procent renhet >95%