Läs mer
Valsartan n-Propyl Impurity, TRC
Produktkod. 30006625
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006625 25 mg
17963755 10 mg
17951215 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30006625 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV09577025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 952652-79-8
Molekylvikt (g/mol) 421.49
Synonym (2S)-2-[Butanoyl[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]amino]-3-methylbutanoic Acid, Valsartan Imp. C (EP), Valsartan Related Compound B (USP)
IUPAC-namn (2S)-2-[butanoyl-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]amino]-3-methylbutanoic acid
LEDER CCCC(=O)N(Cc1ccc(cc1)c2ccccc2c3nnn[nH]3)[C@@H](C(C)C)C(=O)O
Specifikationer
Kemiskt namn eller material (2S)-2-[Butanoyl[(2'-(1H-tetrazol-5-yl)biphenyl-4-yl)methyl]amino]-3-methylbutanoic Acid
Anteckningar om renhetsgrad HPLC
Molekylformel C23 H27 N5 O3
InChI formel InChI=1S/C23H27N5O3/c1-4-7-20(29)28(21(15(2)3)23(30)31)14-16-10-12-17(13-11-16)18-8-5-6-9-19(18)22-24-26-27-25-22/h5-6,8-13,15,21H,4,7,14H2,1-3H3,(H,30,31)(H,24,25,26,27)/t21-/m0/s1
Kvantitet 25 mg
Formel vikt 421.2114
Procent renhet >95%