Läs mer
Vancomycin CDP-1 (>85%), TRC
Produktkod. 30133511
Change view
Kvantitet:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133511 100 mg
30012645 10 mg
30004785 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133511 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV096510100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55598-85-1
Synonym (4S,7R,8R,23R,24S,27S,31aSa,37R,39aR)-45-[[2-O-(3-Amino-2,3,6-trideoxy-3-C-methyl-α-L-lyxo-hexopyranosyl)-β-D-glucopyranosyl]oxy]-11,20-dichloro-8,23,29,31,33-pentahydroxy-7-[[(2R)-4-methyl-2-(methylamino)pentanoyl]amino]-2,6,25,39,40-pentaoxo-1,2,3,4,5,6,7,8,24,25,26,27,37,38,39,39a-hexadecahydro-23H-9,12:19,22-dietheno-24,37-(iminomethano)-14,17:32,36-dimetheno-14H-[1,6,10]oxadiazacycloheptadecino[4,5-m][10,2,16]benzoxadiazacyclotetracosine-4,27-dicarboxylic acid∣23H-9,12:19,22-Dietheno-24,37-(iminomethano)-14,17:32,36-dimetheno-17H-[1,6,10]oxadiazacycloheptadecino[4,5-m][10,2,16]benzoxadiazacyclotetracosine-4,27-dicarboxylic acid, 45-[[2-O-(3-amino-2,3,6-trideoxy-3-C-methyl-α-L-arabino-hexopyranosyl)-β-D-glucopyranosyl]oxy]-11,20-dichloro-1,2,3,4,5,6,7,8,24,25,26,27,37,38,39,39a-hexadecahydro-8,23,29,31,33-pentahydroxy-7-[[4-methyl-2-(methylamino)-1-oxopentyl]amino]-2,6,25,39,40-pentaoxo-∣45-[[2-O-(3-Amino-2,3,6-trideoxy-3-C-methyl-α-L-arabino-hexopyranosyl)-β-D-glucopyranosyl]oxy]-11,20-dichloro-1,2,3,4,5,6,7,8,24,25,26,27,37,38,39,39a-hexadecahydro-8,23,29,31,33-pentahydroxy-7-[[4-methyl-2-(methylamino)-1-oxopentyl]amino]-2,6,25,39,40-pentaoxo-23H-9,12:19,22-dietheno-24,37-(iminomethano)-14,17:32,36-dimetheno-17H-[1,6,10]oxadiazacycloheptadecino[4,5-m][10,2,16]benzoxadiazacyclotetracosine-4,27-dicarboxylic acid∣CDP 1∣CDP-I∣Vancomycin CDP-1∣Vancomycin CDP-I
Specifikationer
Formatera Neat
Molekylformel C66H74Cl2N8O25
Molekylvikt (g/mol) 1450.24, 1450.25
InChI-nyckel OGMARHJAPWBNFA-UHFFFAOYNA-N
LEDER CNC(CC(C)C)C(=O)NC1C(O)C2=CC=C(OC3=CC4=CC(OC5=CC=C(C=C5Cl)C(O)C5NC(=O)C(NC(=O)C4NC(=O)CC(NC1=O)C(O)=O)C1=CC=C(O)C(=C1)C1=C(O)C=C(O)C=C1C(NC5=O)C(O)=O)=C3OC1OC(CO)C(O)C(O)C1OC1CC(C)(N)C(O)C(C)O1)C(Cl)=C2
IUPAC-namn 49-({3-[(4-amino-5-hydroxy-4,6-dimethyloxan-2-yl)oxy]-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl}oxy)-48,50-dichloro-2,18,33,36,38-pentahydroxy-19-[4-methyl-2-(methylamino)pentanamido]-20,24,27,43,45-pentaoxo-7,13-dioxa-21,25,28,42,44-pentaazaoctacyclo[27.14.2.2³,⁶.2¹⁴,¹⁷.1⁸,¹².1³⁰,³⁴.0¹⁰,²⁶.0³⁵,⁴⁰]henpentaconta-3,5,8(49),9,11,14,16,30,32,34(46),35,37,39,47,50-pentadecaene-22,41-dicarboxylic acid
Produkttyp Impurity