Läs mer
Vancomycin Monohydrochloride, TRC
Produktkod. 30456896
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30456896 1 g
1 options
Produktkod. 30456896 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV0965001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Vancomycin Monohydrochloride
CAS 197638-25-8
Molekylformel C66H75Cl2N9O24 • HCl
Kvantitet 1 g
Synonym Lyphocin Monohydrochloride, Meek Monohydrochloride, Vancocin Monohydrochloride, Vancocine Monohydrochloride, Vancocyn Monohydrochloride, Vancor Monohydrochloride
InChI formel InChI=1S/C66H75Cl2N9O24.ClH/c1-23(2)12-34(71-5)58(88)76-49-51(83)26-7-10-38(32(67)14-26)97-40-16-28-17-41(55(40)101-65-56(54(86)53(85)42(22-78)99-65)100-44-21-66(4,70)57(87)24(3)96-44)98-39-11-8-27(15-33(39)68)52(84)50-63(93)75-48(64(94)95)31-18-29(79)19-37(81)45(31)30-13-25(6-9-36(30)80)46(60(90)77-50)74-61(91)47(28)73-59(89)35(20-43(69)82)72-62(49)92;/h6-11,13-19,23-24,34-35,42,44,46-54,56-57,65,71,78-81,83-87H,12,20-22,70H2,1-5H3,(H2,69,82)(H,72,92)(H,73,89)(H,74,91)(H,75,93)(H,76,88)(H,77,90)(H,94,95);1H/t24-,34+,35-,42?,44-,46?,47+,48-,49+,50-,51+,52+,53?,54?,56+,57+,65-,66-;/m0./s1
LEDER [H][C@@]([C@](NC([C@@](NC)([H])CC(C)C)=O)([H])C1=O)(O)C(C=C2Cl)=CC=C2OC3=C(O[C@@H]4OC(CO)C(O)C(O)[C@H]4O[C@@H]5O[C@@H](C)[C@@H](O)[C@](N)(C)C5)C(OC6=CC=C([C@]7(O)[H])C=C6Cl)=CC([C@]([H])(C(NC8([H])C(N[C@]7([H])C(N[C@]([H])(C(O)=O)C(C=C(O)C=C9O)=C9C%10=C(O)C=CC8=C%10)=O)=O)=O)NC([C@@](CC(N)=O)([H])N1)=O)=C3.Cl
Molekylvikt (g/mol) 1449.253646
Formel vikt 1483.4069