Läs mer
Vanillin-13C6, TRC
Produktkod. 17960965
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17960965 1 mg
17900955 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17960965 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV0975031MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 201595-58-6
Molekylformel 13C6 C2 H8 O3
Molekylvikt (g/mol) 158.1
Synonym Benzaldehyde-1,2,3,4,5,6-13C6, 4-hydroxy-3-methoxy- (9CI), 4-Hydroxy-3-methoxybenzaldehyde-1,2,3,4,5,6-13C6, 4-Hydroxy-3-methoxybenzaldehyde-13C6, 4-Hydroxy-3-methoxybenzaldehyde-ring-13C6, 4-Hydroxy-3-methoxybenzaldehyde-13C6 (phenyl-13C6), Vanillin-13C6
IUPAC-namn 4-hydroxy-3-methoxy-(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-triene-1-carbaldehyde
LEDER CO[13c]1[13cH][13c]([13cH][13cH][13c]1O)C=O
Specifikationer
Kemiskt namn eller material Vanillin (13C6)
Kvantitet 1 mg
InChI formel InChI=1S/C8H8O3/c1-11-8-4-6(5-9)2-3-7(8)10/h2-5,10H,1H3/i2+1,3+1,4+1,6+1,7+1,8+1
Formel vikt 158.0675