missing translation for 'onlineSavingsMsg'
Learn More

Vanillin, 99%, MP Biomedicals™

Catalog Number p-4404907
Click to view available options
Quantity:
100 g
500 g
1 kg
This item is not returnable. View return policy
121-33-5
C8H8O3
152.15
MFCD00006942,MFCD08702848
MWOOGOJBHIARFG-UHFFFAOYSA-N
vanillin, vanillaldehyde, vanillic aldehyde, vanilla, p-vanillin, 2-methoxy-4-formylphenol, vanilline, lioxin, 4-hydroxy-m-anisaldehyde, 3-methoxy-4-hydroxybenzaldehyde
1183
CHEBI:18346
4-hydroxy-3-methoxybenzaldehyde
COC1=CC(C=O)=CC=C1O
This item is not returnable. View return policy
121-33-5
C8H8O3
152.15
MFCD00006942,MFCD08702848
MWOOGOJBHIARFG-UHFFFAOYSA-N
vanillin, vanillaldehyde, vanillic aldehyde, vanilla, p-vanillin, 2-methoxy-4-formylphenol, vanilline, lioxin, 4-hydroxy-m-anisaldehyde, 3-methoxy-4-hydroxybenzaldehyde
1183
CHEBI:18346
4-hydroxy-3-methoxybenzaldehyde
COC1=CC(C=O)=CC=C1O
Color White
Density 1.056g/mL
Assay Percent Range 99%
Quantity 100 g
Formula Weight 152.1
Physical Form Crystalline Powder
Chemical Name or Material Vanillin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'