Learn More
Thermo Scientific™ Vanquish™ Split Sampler Sample Loops
Product Code. 16371298 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Vanquish Split Sampler HT and FT
Sample Capacity:
10 μL
Biocompatible Version:
Yes
Unit Size:
Each
Product Code. For Use With (Equipment) Sample Capacity Biocompatible Version Unit Size
16371298 Vanquish Split Sampler HT and FT 10 μL Yes Each
1 options
Product Code. 16371298 Supplier Thermo Scientific™ Supplier No. 6850.1915
 Restricted Item
Beräknad leverans: 14-12-2026
Thermo Scientific™ Vanquish™ Split Sampler Sample Loops come in a variety of sizes to fit your application requirements..
Automate sample injection into your Thermo Scientific™ Vanquish™ HPLC or UHPLC system with Thermo Scientific™ Vanquish™ Split Sampler Sample Loops. These sample loops are available in 10, 25, 100, 250, 1000 μL sizes to meet your application requirements. Thermo Scientific™ Viper™ Fittings are included with our sample loops to make them easy to install with leak-free connections.
Vanquish Split Sampler Sample Loops use Viper Fittings, which have a ferrule-free tightening design, for virtually zero-dead-volume connections.
Specifications
Type Sample Loop
Sample Capacity 10 μL
Biocompatible Version Yes
For Use With (Equipment) Vanquish Split Sampler HT and FT