missing translation for 'onlineSavingsMsg'
Learn More

VAP1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14365-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16892974

  • 1656.73 SEK / 20µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Copper amine oxidases catalyze the oxidative conversion of amines to aldehydes in the presence of copper and quinone cofactor. The product is a major protein on the adipocyte plasma membrane. It has adhesive properties and also has functional monoamine oxidase activity. A pseudogene for this gene has been described and is located approximately 9-kb downstream.
Specifications

Specifications

VAP1
Polyclonal
Unconjugated
AOC3
AOC3, Copper amine oxidase, HPAO, Membrane primary amine oxidase, SSAO, VAP 1, VAP1, Vascular adhesion protein 1
Rabbit
Antigen Affinity Chromatography
RUO
8639
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q16853
AOC3
VAP1 Fusion Protein Ag5683
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers