missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ VapLock™ Adapters for Exhaust Filters, NPT(F)
VapLock™ Adapters for Exhaust Filters, NPT(F) feature various adapter configurations allowing quick and easy connection to emission sources
POA
Specifications
For Use With (Application) | Solvent Safety |
---|---|
For Use With (Equipment) | VapLock Exhaust Filters |
Material | Black Acetal |
Size | NPT(M) x NPT(F), adjustable |
Description
- Fit VapLock exhaust filters, which have 1/4 in NPT(F) ports
- May require additional adapters to fit your container
- Adjustable for flexible positioning
Warranty
None
Specifications
Solvent Safety | |
VapLock Exhaust Filters | |
Black Acetal | |
NPT(M) x NPT(F), adjustable |