Learn More
Cole-Parmer™ VapLock™ Breakthrough Color Indicator for Exhaust Filters
Use VapLock™ Breakthrough Color Indicator for Exhaust Filters to detect end-of-service life
POA
Specifications
Type | Breakthrough Indicator for Exhaust Filters |
---|---|
For Use With (Application) | Solvent Safety |
For Use With (Equipment) | VapLock Exhaust Filters |
Material | Acrylic |
Description
- Changes color when exposed to organic solvent vapors
- Optically clear acrylic housing for easy view
Warranty
None
Specifications
Breakthrough Indicator for Exhaust Filters | |
Solvent Safety | |
VapLock Exhaust Filters | |
Acrylic |