Läs mer
Vardenafil Dihydrochloride, LoGiCal
Produktkod. 30463058
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30463058 10 mg
1 options
Produktkod. 30463058 Leverantör LoGiCal Leverantörsnummer LGCFOR1619.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Vardenafil dihydrochloride
CAS 224789-15-5
Analyt- eller komponentnamn Vardenafil dihydrochloride
Frakt skick Room Temperature
Molekylformel C23 H32 N6 O4 S . 2 Cl H
Certifikat och efterlevnad ISO 9001
Synonym Imidazo[5,1-f][1,2,4]triazin-4(1H)-one, 2-[2-ethoxy-5-[(4-ethyl-1-piperazinyl)sulfonyl]phenyl]-5-methyl-7-propyl-, hydrochloride (1:2), Piperazine, 1-[[3-(1,4-dihydro-5-methyl-4-oxo-7-propylimidazo[5,1-f][1,2,4]triazin-2-yl)-4-ethoxyphenyl]sulfonyl]-4-ethyl-, dihydrochloride (9CI), Vardenafil dihydrochloride
InChI formel InChI=1S/C23H32N6O4S.2ClH/c1-5-8-20-24-16(4)21-23(30)25-22(26-29(20)21)18-15-17(9-10-19(18)33-7-3)34(31,32)28-13-11-27(6-2)12-14-28;;/h9-10,15H,5-8,11-14H2,1-4H3,(H,25,26,30);2*1H
Lösningstyp Neat
LEDER Cl.Cl.CCCc1nc(C)c2C(=O)NC(=Nn12)c3cc(ccc3OCC)S(=O)(=O)N4CCN(CC)CC4
IUPAC-namn 2-[2-ethoxy-5-(4-ethylpiperazin-1-yl)sulfonylphenyl]-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one;dihydrochloride
Industry Type Clinical Testing
Molekylvikt (g/mol) 561.52
Kvantitet 10 mg
Formel vikt 560.1739 g/mol