missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vardenafil Dihydrochloride, Mikromol™
Description
Vardenafil Dihydrochloride
Specifications
Specifications
| Kemiskt namn eller material | Vardenafil dihydrochloride |
| CAS | 224789-15-5 |
| Analyt- eller komponentnamn | Vardenafil dihydrochloride |
| Frakt skick | Room Temperature |
| Molekylformel | C23 H32 N6 O4 S . 2 Cl H |
| Certifikat och efterlevnad | ISO 17034 |
| Synonym | Imidazo[5,1-f][1,2,4]triazin-4(1H)-one, 2-[2-ethoxy-5-[(4-ethyl-1-piperazinyl)sulfonyl]phenyl]-5-methyl-7-propyl-, hydrochloride (1:2), Piperazine, 1-[[3-(1,4-dihydro-5-methyl-4-oxo-7-propylimidazo[5,1-f][1,2,4]triazin-2-yl)-4-ethoxyphenyl]sulfonyl]-4-ethyl-, dihydrochloride (9CI), Vardenafil dihydrochloride |
| InChI formel | InChI=1S/C23H32N6O4S.2ClH/c1-5-8-20-24-16(4)21-23(30)25-22(26-29(20)21)18-15-17(9-10-19(18)33-7-3)34(31,32)28-13-11-27(6-2)12-14-28;;/h9-10,15H,5-8,11-14H2,1-4H3,(H,25,26,30);2*1H |
| LEDER | Cl.Cl.CCCc1nc(C)c2C(=O)NC(=Nn12)c3cc(ccc3OCC)S(=O)(=O)N4CCN(CC)CC4 |
| IUPAC-namn | 2-[2-ethoxy-5-(4-ethylpiperazin-1-yl)sulfonylphenyl]-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one;dihydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?