Läs mer
Vardenafil hydrochloride, MedChemExpress
Produktkod. 16498306
Encompass_Preferred
Change view
Kvantitet:
100 mg
200 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16498306 100 mg
16401426 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16498306 Leverantör Medchem Express Leverantörsnummer HYB0442A
MedChemExpress Vardenafil hydrochloride is a selective and orally active inhibitor of phosphodiesterase-5 (PDE5), with an IC50 of 0.7 nM.
Vardenafil hydrochloride shows inhibitory towards PDE1 (180 nM), PDE6 (11 nM), PDE2, PDE3, and PDE4 (>1000 nM). Vardenafil hydrochloride competitively inhibits cyclic guanosine monophosphate (cGMP) hydrolysis and thus increases cGMP levels. Vardenafil hydrochloride can be used for the research like erectile dysfunction, hepatitis, diabetes[1]-[6].
Specifikationer
Kemiskt namn eller material Vardenafil hydrochloride
CAS 224785-91-5
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C23H33ClN6O4S
Kvantitet 100 mg
Löslighetsinformation DMSO : 100 mg/mL (190.45 mM; Need ultrasonic) ∣H2O : ≥ 100 mg/mL (190.45 mM)
LEDER CCCC1=NC(C)=C2N1N=C(C(C=C(S(=O)(N3CCN(CC)CC3)=O)C=C4)=C4OCC)NC2=O.Cl
Molekylvikt (g/mol) 525.06
Formel vikt 525.06
Procent renhet 99.48%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation