Läs mer
Thermo Scientific™ Vari-Mix™ and Speci-Mix™ Test Tube Rockers
Obtain uniform specimen suspension with the smooth rocking action of these compact, precision-controlled platform mixers.
Beskrivning
Beskrivning
Obtain uniform specimen suspension with the smooth rocking action of Thermo Scientific™ Vari-Mix™ and Speci-Mix™ Test Tube Rockers. Compact and precision-controlled platform mixers, all models hold 10mm to 30mm diameter tubes and are ideal for blood collection tubes.
  • Choose from fixed or adjustable speeds
  • Reversible pad provides one side (with lip to prevent sliding) for capped tubes 110mm (4.33″) long; other side for longer tubes
  • Autoclavable white silicone rubber pad enhances viewing
  • Can be used in a noncondensing incubator or coldroom with ambient operating conditions of 4°C to 40°C (39.2°F; to 104°F) and 20% to 80% RH noncondensing
  • Includes white silicon rubber friction grip pad for holding tubes in place
Warranty: 90 days
Filmer
SDS
Dokument
Dokument
Produktspecifikationer
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.