missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ GC Inlet Liner Large Volume Injection (Lvi) Sintered Glass For Varian 1078/1079 1.8/3.4mm I.D.
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan Scientific and Medical™ 09224525
Additional Details : Weight : 0.10000kg
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Varian instruments
Specifications
Large Volume Injection (LVI) Liner | |
54 mm | |
5 mm | |
Sintered Glass |
25/Pk. | |
1.8, 3.4 mm | |
Varian GC Instruments |