Learn More
Thermo Scientific™ Vascular Endothelial Growth Factor-165, human
Brand:  Thermo Scientific™ J67388.&E
Additional Details : Weight : 0.00001kg
 View more versions of this product
Product Code. 15460737
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
38.2 kd
Plastic bottle
Lyophilized
2μg
Store at -20°C
Product Suggestions
Product Suggestions
Videos
SDS Documents