missing translation for 'onlineSavingsMsg'
Läs mer

Vat Orange 3 (Technical Grade), TRC

Produktkod. 30456317
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
2.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30456317 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456317 Supplier Toronto Research Chemicals Supplier No. TRCV9950382.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Vat Orange 3 (Technical Grade)
CAS 4378-61-4
Molekylformel C22H8Br2O2
Kvantitet 2.5 g
InChI formel InChI=1S/C22H8Br2O2/c23-15-7-14-19-17-9(15)3-1-5-11(17)21(25)13-8-16(24)10-4-2-6-12(22(14)26)18(10)20(13)19/h1-8H
LEDER Brc1cc2C(=O)c3cccc4c(Br)cc5C(=O)c6cccc1c6c2c5c34
Molekylvikt (g/mol) 464.11
Formel vikt 461.8891
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.