Learn More
VAT1L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560605
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DNPPKTPLVP GFECSGIVEA LGDSVKGYEI GDRVMAFVNY NAWAEVVCTP VEFVYKIPDD MSFSEAAAFP MNFVTAYVML FEVANL Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
VAT1L is a protein coding gene.Specifications
VAT1L | |
Polyclonal | |
Unconjugated | |
VAT1L | |
9430073I07; AI427515; Kiaa1576; mKIAA1576; Synaptic vesicle membrane protein VAT-1 homolog-like; Vat1l; vesicle amine transport 1-like; vesicle amine transport protein 1 homolog (T. californica)-like; vesicle amine transport protein 1 homolog-like (T. californica) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57687 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HCJ6 | |
VAT1L | |
Recombinant protein corresponding to Human VAT1L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only