Läs mer
Invitrogen™ VCAM-1 Monoclonal Antibody (1.4C3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA511447
Beskrivning
MA5-11447 targets CD106 in IHC (P, F) and ICC/IF applications and shows reactivity with Human samples. The MA5-11447 immunogen is stimulated human umbilical vein endothelial cells (HUVEC). MA5-11447 has been successfully used in immunohistochemistry analysis of VCAM1 in mouse E16.5 frozen lung sections.
VCAM-1 is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. The protein has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases.
Specifikationer
| VCAM-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.05% BSA and 0.05% sodium azide; pH 7.4 | |
| P19320, P29533 | |
| VCAM1 | |
| Stimulated human umbilical vein endothelial cells (HUVEC). | |
| 500 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1.4C3 | |
| Unconjugated | |
| VCAM1 | |
| adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 22329, 7412 | |
| 4°C | |
| Liquid |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.