missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ VCX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561942
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: MSPKPRASGP PAKATEAGKR KSSSQ Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 50%.
This gene belongs to the VCX/Y gene family, which has multiple members on both X and Y chromosomes, and all are expressed exclusively in male germ cells. The X-linked members are clustered on chromosome Xp22 and Y-linked members are two identical copies of the gene within a palindromic region on Yq11. The family members share a high degree of sequence identity, with the exception that a 30-bp unit is tandemly repeated in X-linked members but occurs only once in Y-linked members. The VCX gene cluster is polymorphic in terms of copy number; different individuals may have a different number of VCX genes. VCX/Y genes encode small and highly charged proteins of unknown function. The presence of a putative bipartite nuclear localization signal suggests that VCX/Y members are nuclear proteins. This gene contains 10 repeats of the 30-bp unit.
Specifikationer
| VCX | |
| Polyclonal | |
| Unconjugated | |
| VCX | |
| Variable charge protein on X with ten repeats; variable charge X-linked protein 1; variable charge, X chromosome; variable charge, X-linked; variably charged protein X-B1; VCX; VCX1; VCX10R; VCX-10r; VCXB1; VCX-B1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26609 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H320 | |
| VCX | |
| Recombinant protein corresponding to Human VCX. Recombinant protein control fragment (Product #RP-104447). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering