missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ VDAC1 (Human) Recombinant Protein
Human VDAC1 partial ORF ( AAH08482, 1 a.a. - 100 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00007416-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MAVPPTYADLGKSARDVFTKGYGFGLIKLDLKTKSENGLEFTSSGSANTETTKVTGSLETKYRWTEYGLTFTEKWNTDNTLGTEITVEDQLARGLKLTFD
Specifications
AAH08482 | |
voltage-dependent anion channel 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
VDAC1 | |
GST |
7416 | |
Wheat germ expression system | |
10 μg | |
MGC111064, PORIN, PORIN-31-HL | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |